Skip to Content
Merck
Siliceous sand CP

Siliceous sand

Siliceous sand CP
Synonym(s):
Sand, white quartz, Quartz, Silicon dioxide
Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
EC Number:

Products

Brand
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing