Skip to Content
Merck
Perphenazine sulfoxide solution 1 mg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Perphenazine sulfoxide solution

Perphenazine sulfoxide solution 1 mg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®
Synonym(s):
Perphenazine sulfoxide, 4-[3-(2-Chlorophenothiazin-10-yl)propyl]-1-piperazineethanol sulfoxide1-Piperazineethanol sulfoxide
Empirical Formula (Hill Notation):
C21H26ClN3O2S
CAS Number:
Molecular Weight:
419.97

Products

Brand
Formula Weight
Manufacturer
Format
Quality Segment
Package Size
Special Grade