JP
EN
Applications
Products
Services
Support
Home
Search Results
Moisture in methanol, 325 mg/kg
(1)
Moisture in methanol, 325 mg/kg
Synonyms:
Methyl alcohol, Methanol
Linear Formula:
CH
3
OH
CAS Number:
67-56-1
Molecular Weight:
32.04
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
NISTRM8510
Moisture in methanol, 325 mg/kg, NIST
®
SRM
®
8510
Expand
View Pricing