Merck
Maleic acid SAJ special grade, ≥99.0%

Maleic acid

Maleic acid SAJ special grade, ≥99.0%
Synonyms:
cis-Butenedioic acid, Toxilic acid
Linear Formula:
HO2CCH=CHCO2H
Molecular Weight:
116.07
EC Number:

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Maleic acid, ReagentPlus®, ≥99% (HPLC)
Maleic acid, Standard for quantitative NMR, TraceCERT®
Maleic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Maleic acid, for synthesis
Maleic acid, United States Pharmacopeia (USP) Reference Standard
Maleic acid, electronic grade, ≥99.97% trace metals basis, water <=350 ppm
Maleic acid, ≥99.5% (HPLC), <=0.5% water
Maleic acid, phyproof® Reference Substance
Maleic acid
Maleic acid, European Pharmacopoeia (EP) Reference Standard
Maleic acid, SAJ special grade, ≥99.0%