(-)-Levamisole-D5 hydrochloride solution
Empirical Formula (Hill Notation):
C11H7D5N2S · HCl
CAS Number:
Molecular Weight:
245.78
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing