Skip to Content
Merck
Iron(III) phosphate for analysis calcined (max. 0.001% SO₄) EMSURE®

Iron(III) phosphate

Iron(III) phosphate for analysis calcined (max. 0.001% SO₄) EMSURE®
Synonym(s):
Iron(III) phosphate
Empirical Formula (Hill Notation):
FeO4P
CAS Number:
Molecular Weight:
150.82

Products

Product Category
Brand
Sustainability
Feature
Formula Weight
Physical Form
Product Line
Chemical Composition
Quality Segment