JP
EN
Applications
Products
Services
Support
Home
Search Results
Hydrogen chloride – methanol solution
(1)
Hydrogen chloride – methanol solution
Synonyms:
Hydrochloric acid in methanol
Linear Formula:
HCl · CH
3
OH
CAS Number:
132228-87-6
Molecular Weight:
36.46
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
17935
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
™
Expand
View Pricing