JP
EN
Applications
Products
Services
Support
Home
Search Results
Hydrogen chloride – ethanol solution
(3)
Hydrogen chloride – ethanol solution
Synonyms:
Hydrochloric acid in ethanol
Linear Formula:
HCl · CH
3
CH
2
OH
CAS Number:
7647-01-0
Molecular Weight:
36.46
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
17934
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
™
Expand
View Pricing
13-1800
Hydrogen chloride – ethanol solution, 3% in ethanol
Expand
View Pricing
13-1790
Hydrogen chloride – ethanol solution, 0.1 M in ethanol
Expand
View Pricing