Merck
Hydrogen chloride – 2-propanol solution ~1.25 M HCl (T), for GC derivatization, LiChropur™

Hydrogen chloride – 2-propanol solution

Hydrogen chloride – 2-propanol solution ~1.25 M HCl (T), for GC derivatization, LiChropur™
Synonyms:
Hydrochloric acid in 2-propanol
Linear Formula:
HCl · CH3CH(OH)CH3
Molecular Weight:
36.46

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur