JP
EN
Applications
Products
Services
Support
Home
Search Results
Hydrochloric acid fuming 37%
(2)
Hydrochloric acid fuming 37%
Synonyms:
Hydrochloric acid
Empirical Formula (Hill Notation):
HCl
CAS Number:
7647-01-0
Molecular Weight:
36.46
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
1.00317
Hydrochloric acid fuming 37%, for analysis EMSURE
®
ACS,ISO,Reag. Ph Eur
Expand
View Pricing
1.13386
Hydrochloric acid fuming 37%, for analysis max. 0.001 ppm Hg EMSURE
®
Expand
View Pricing