Merck
Hydrochloric acid 30% Ultrapur

Hydrochloric acid 30%

Hydrochloric acid 30% Ultrapur
Synonyms:
Hydrogen chloride solution, Hydrochloric acid solution
Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Hydrochloric acid 30%, Suprapur®
Hydrochloric acid 30%, Ultrapur