JP
EN
Applications
Products
Services
Support
Home
Search Results
Formic acid solution
(1)
Formic acid solution
Linear Formula:
HCOOH
CAS Number:
64-18-6
Molecular Weight:
46.03
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
06473
Formic acid solution, BioUltra, 1.0 M in H
2
O
Expand
View Pricing