JP
EN
Applications
Products
Services
Support
Home
Search Results
Dextran 40
(1)
Dextran 40
Synonyms:
Dextran
Linear Formula:
[C
6
H
10
O
5
]
n
CAS Number:
9004-54-0
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
1179708
Dextran 40, United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing