
Bismuth subcarbonate

Synonym(s):
Bismuth(III) carbonate basic, Bismuth subcarbonate
Linear Formula:
(BiO)2CO3
CAS Number:
Molecular Weight:
509.97
Products
Formula Weight
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing