Skip to Content
Merck
Agar Strips SDX-y Sabouraud Dextrose Agar, Yeasts and molds, y-irradiated, HYCON®

Agar Strips SDX-y

Agar Strips SDX-y Sabouraud Dextrose Agar, Yeasts and molds, y-irradiated, HYCON®
Synonym(s):
Agar Strips

Products

Product Category
Brand
Physical Form
Compatibility
Quality Segment
Application
Microorganism Suitability
Package Size
Sterility
Sterilization Method
Showing 1-1 of 1