Skip to Content
Merck
Activated carbon SAJ first grade

Activated carbon

Activated carbon SAJ first grade
Synonym(s):
Activated charcoal, Charcoal activated
Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Showing 1-2 of 2
Compare
Product Number
Description
Pricing