Skip to Content
Merck
1-Methyl-2-pyrrolidinone puriss. p.a., ≥99.0% (GC)

1-Methyl-2-pyrrolidinone

1-Methyl-2-pyrrolidinone puriss. p.a., ≥99.0% (GC)
Synonym(s):
1-Methyl-2-pyrrolidone, N-Methyl-2-pyrrolidone, NMP, 1-Methyl-2-pyrrolidinone, 1-Methyl-2-pyrrolidinone solution
Empirical Formula (Hill Notation):
C5H9NO
CAS Number:
Molecular Weight:
99.13
EC Number:

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique