Merck
4-(3,6-Dimethyl-3-heptyl)phenol monoethoxylate-ring-13C6 solution 10 μg/mL in acetone, analytical standard

4-(3,6-Dimethyl-3-heptyl)phenol monoethoxylate-ring-13C6 solution

4-(3,6-Dimethyl-3-heptyl)phenol monoethoxylate-ring-13C6 solution 10 μg/mL in acetone, analytical standard
Synonyms:
2-[4-(1-Ethyl-1,4-dimethylpentyl)phenoxy]ethanol-13C6, 2-[4-(3,6-Dimethyl-3-heptyl)phenoxy]ethanol-13C6, 3,6,3-Nonylphenol-monoethoxylate-13C6, 363-NP1EO-13C6
Empirical Formula (Hill Notation):
13C6C11H28O2
CAS Number:
Molecular Weight:
270.36
EC Number:

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
4-(3,6-Dimethyl-3-heptyl)phenol monoethoxylate-ring-13C6 solution, 10 μg/mL in acetone, analytical standard