Skip to Content
Merck
Colchicine ≥95% (HPLC), powder

Colchicine

Colchicine ≥95% (HPLC), powder
Synonym(s):
(S)-N-(5,6,7,9-Tetrahydro-1,2,3,10-tetramethoxy-9-oxobenzo[a]heptalen-7-yl)acetamide, Colchicine, Colchicum autumnale
Empirical Formula (Hill Notation):
C22H25NO6
CAS Number:
Molecular Weight:
399.44

Products

Shipping
Product Category
Brand
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Research Area
Quality Segment
Application
Technique