Skip to Content
Merck
L-Tyrosine reagent grade, ≥98% (HPLC)

L-Tyrosine

L-Tyrosine reagent grade, ≥98% (HPLC)
Synonym(s):
(S)-2-Amino-3-(4-hydroxyphenyl)propionic acid, 3-(4-Hydroxyphenyl)-L-alanine, L-Tyrosine, (S)-(-)-Tyrosine
Linear Formula:
4-(HO)C6H4CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
181.19

Products

Shipping
Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Optical Isomer
Amino Acid
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing