Heparin sodium salt from porcine intestinal mucosa
Synonym(s):
Heparin sodium salt from porcine intestinal mucosa, Heparin, Sodium Salt, Porcine Intestinal Mucosa
CAS Number:
Products
Shipping
Product Category
Brand
Biological Source
Color
Feature
Physical Form
Product Line
Research Area
Tissue
Quality Segment
Application
Package Size
Technique
Showing 1-7 of 7
Compare
Product Number
Description
Pricing