Merck
Cryo Storage
Lipids and fatty acids standards for testing industries.
We offer a wide range of cryogenic tools and supplies including freezer tubes, vials, racks, labels, markers, storage boxes, and gloves. Our cryo products are suitable for use with autoclaves, freezers, handling liquid nitrogen, oxygen or helium from cryogenic atmospheres, cold rooms, blast freezers, filling of cryogenic liquids, and dry ice handling. We provide a diverse range of cryo supplies for research and clinical applications including genetic, biotech and pharmaceutical laboratories, fertility clinics, and other industries.
Cryo-gloves®
Choose from our selection of various sizes, water resistant cryo gloves, to provide you with excellent thermal protection, flexibility, and dexterity when working in extreme environments (-160 to 150°C).
  • Style provides additional warmth and comfort
  • Oversized for better insulation
  • Resistant to solvents/chemicals
  • Breathable and static-free
  • Machine washable/dryable inner lining
  • Suitable for Class 1000 clean room use
  • Asbestos-free
Storage racks and freezer boxes
We carry a wide range of durable racks and freezer boxes for your cryo material organization. Choose from popular brands including Arctic Squares®, BRAND®, Corning®, Nalgene®, Nunc®, TrueNorth®, and many others.
  • Racks are stackable, with alphanumerical positions.
  • Compositions range from cardboard to stainless steel.
  • Available in different and colors and configurations.
Cryo labels and markers
We offer a full range of durable labels and markers from Diversified Biotech including Cryo-tags™, Cryo-babies™, Tough Tags™, Tough-Spots™, and others. These labels provide colored, chemically inert, temperature resistant tagging to most plastics, glass, and metals without any degradation. The selection also includes specially designed laser cryo-tags for ease of printing.
Cryo vials and Cryo tubes
We offer an extensive range of cryogenic tubes and vials designed to protect and store biological samples, human or animal cells, at low temperatures in the gas phase of liquid nitrogen. Our non-cytotoxic and pyrogen-free cryogenic storage tubes are sterile, temperature resistant, and free-standing. We guarantee external threading and absolute gas tightness in our vials without the use of a silicone gasket.
Other cryo accessories
From chillDown beakers to cryo forceps, we offer the tools required for safe and efficient production in the cryo-environment. Our ChillDown polyethylene beakers are ideal for rapid cooling of agarose solutions. The cryopreservative forceps allow easy manipulation of cryopreservation straws, vials, and other small objects.
Related Video