Custom Peptides
- High quality peptide synthesis using FMOC and/or BOC chemistries
- Sequence length: 2-40mer
- Amount: up to 1 gram
- Purity Levels: Unpurified, 70%, 80%, 95%
- Comprehensive Modification offering including, FRET, C-terminal aldehyde, phosphorylation, multiple disulfide bonds, etc.
- Format: Supplied lyophilized
- QC Analysis: All peptides by HPLC and MS; additional analysis available upon request
Peptide Library Design
- Design overlapping peptide fragment libraries, alanine scanning libraries, truncation libraries and positional scanning libraries
- Set custom sequence parameters (peptide length and amino acid offset)
- Format the sequence output
- Export the custom peptide library sequences (MS Word, Excel, etc.)
PEPscreen®: Custom Peptide Libraries
- Suitable for robotic production of peptide microarrays
- Screen mulitple proteotyptic peptides and rapidly characterize and select mass spec compatible peptides
- Get results quick with delivery of your peptide library in less than 7 business days
- Sequence Length: 6 to 20 amino acids
- Amount: 0.5-2mg or 2-5mg
- Modifications: Comprehensive offering including phosphorylation, biotin, fluorescein, stable isotopes, etc.
- Format: Supplied dry in a 96 well tube rack
- Options: Normalization and aliquoting (stock and copy libraries)
- Minimum library size of 24 peptides
AQUA™ Peptides
- Accurately quantitate low abundance proteins
- Measure site-specific phosphorylation states
- Validate gene silencing at the protein level
- Custom peptides are available either without stable isotope labeling, or with stable isotope labeling
- Stable-isotope labeled AQUA™ Peptide seqeunces contain one isotopically labeled amino acid
- Sequence Length: 5 to 30 amino acids
- Amount: 5 x 1 nmole
- Purity: >95% by RP-HPLC
- Modifications: phosphorylation (Ser, Thr, Tyr), carboxymethylated Cys, carbamidomethylated Cys, hydroxyproline, N-terminal biotin (please inquire regarding other modifications)
- Quantification by amino acid analysis (AAA)
- Format: Supplied dry