Merck
All Photos(1)

861304

Supelco

EPA 8270 Calibration Mix 6

certified reference material, 1000 μg/mL each component in dichloromethane

grade

certified reference material
TraceCERT®

Quality Level

Agency

EPA 8270

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 2 mL

concentration

1000 μg/mL each component in dichloromethane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

-10 to -25°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Analyte

Description

    Acetophenone
    4-Aminobiphenyl
    m-Cresol
    2,6-Dichlorophenol
    p-Dimethylaminoazobenzene
    7,12-Dimethylbenz[a]anthracene
    1,3-Dinitrobenzene
    Dinoseb
    Diphenylamine
    Ethyl methanesulfonate
    N-(2-Fluorenyl)acetamide
    Hexachloropropene
    Isosafrol
    Methapyrilene HCl
    3-Methylcholanthrene
    Methyl methanesulfonate
    2-Methyl-5-nitroaniline
    1-Naphthylamine
    2-Naphthylamine
    4-Nitroquinoline N-oxide
    N-Nitrosodibutylamine
    N-Nitrosodiethylamine
    N-Nitrosomethylethylamine
    Nitrosomorpholine
    1-Nitrosopiperidine
    1-Nitrosopyrrolidine
    Pentachlorobenzene
    Pentachloroethane
    Pentachloronitrobenzene
    Phenacetin
    2-Picoline
    Safrole
    1,2,4,5-Tetrachlorobenzene
    2,3,4,6-Tetrachlorophenol
    o-Toluidine
    1,3,5-Trinitrobenzene
    See All (36)

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Carc. 1A - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service