29633-U
cone tip stainless steel needle
23 ga
color (light orange)
needle type R (removable needle)
point style 1
pkg of 1 ea
SGE 000492
50 mm × 0.63 mm
0.5 μL
Looking for similar products? Visit Product Comparison Guide
13 - Non Combustible Solids
WGK 3
Not applicable
Not applicable
Enter Lot Number to search for Certificate of Analysis (COA).
Enter Lot Number to search for Certificate of Origin (COO).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service