All Photos(1)
29622-U
SGE autosampler syringes, fixed needle, for CTC/LEAP
5F-C/F-5/0.63, Slim line, fixed needle, volume 5 μL, needle size 23 ga, needle L × O.D. 50 mm × 0.63 mm
NACRES:
NB.03
material
cone tip stainless steel needle
needle size
23 ga
description
color (lime)
feature
needle type F (cemented needle)
point style 1
packaging
pkg of 1 ea
manufacturer/tradename
SGE 001981
needle L × O.D.
50 mm × 0.63 mm
volume
5 μL
Related Categories
Certificate of Analysis
Enter Lot Number to search for Certificate of Analysis (COA).
Certificate of Origin
Enter Lot Number to search for Certificate of Origin (COO).