Merck
All Photos(1)

24934

Supelco

Purge/Trap E

for use with OI Analytical Eclipse 4660, 4560

Synonym(s):
PURGE TRAP E, O.I. ANALYTICAL 4560

application(s)

environmental

compatibility

for use with OI Analytical Eclipse 4660, 4560

Looking for similar products? Visit Product Comparison Guide

Related Categories

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service