All Photos(1)
23984
Headspace/Soil-Gas Syringe
5MDR-HS-V, volume 5 mL, needle size 23 ga, needle L × O.D. 50 mm × 0.63 mm
Synonym(s):
SYRINGE SGE 5MAX-HSV, EA
material
bevel tip needle
glass syringe
needle size
23 ga
feature
Syringe supplied with probe and needle
packaging
pkg of 1 ea
manufacturer/tradename
SGE 008775
needle L × O.D.
50 mm × 0.63 mm
volume
5 mL
General description
  • Fitted with valve and special Luer probe with side hole
  • Needle valve in hub allows gas sample to be stored in syringe during transfer to lab
  • Recommended for soil headspace extraction
  • Probe is removable and replaced with needle during injection in GC
Certificate of Analysis
Enter Lot Number to search for Certificate of Analysis (COA).
Certificate of Origin
Enter Lot Number to search for Certificate of Origin (COO).