Merck
All Photos(1)

21143

Supelco

Purge/Trap I Vocarb® 4000

for use with OI Analytical 4460

Synonym(s):
Vocarb 4000 Purge Trap 0.I.Analytical

application(s)

environmental

compatibility

for use with OI Analytical 4460

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

Vocarb is a registered trademark of Siemens Industry, Inc.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service