SPE075

Supelco

Free Liquids in Paint - PT

Proficiency Testing Material

NACRES:
NA.24

Quality Level

packaging

ampule of 1 × 100 mL

matrix

latex paint

Featured Industry

Environmental

format

matrix material

General description

Free Liquids in Paint proficiency testing material is produced in accordance with ISO/IEC 17043:2010. The PT portfolio allows our test components to offer significant performance data to monitor and prove competency to customers, accreditation bodies or both.

Supelco® is a global proficiency testing (PT) provider that manufactures best-in-class environmental and pharmaceutical PT products. We are the world leader in the manufacturing of real-world soil and sediment PT products.

Type of PT Scheme:
Solids/Soils Hazardous Waste

Matrix: Soil

Analyte/s: Values of analytes vary lot to lot.

All information regarding the use of Free Liquids in Paint proficiency testing standard can be found in the reporting packet which will be available to download at PT Portal.

Application

Free Liquids in paint is an environmental proficiency testing material, which finds applications in the testing of free liquids in paint samples.

Have any questions? Explore our FAQs
For more information on this product, please contact our PT Service Team.

Features and Benefits

We have a vast portfolio of environmental PT schemes with "fit-for-purpose" samples to help you meet the needs of the laboratory.

Our proficiency testing (PT) program is a benchmark to:

  • provide staff with an insight into their laboratory′s performance, to verify the accuracy and reliability of their testing process
  • provide a comparison of performance with that of other laboratories, nationally and internationally
  • test and educate staff, provide a better understanding of the impact of incorrect results and identify areas where there may be problems
  • provide an independent evaluation of laboratory performance
  • demonstrate to clients, colleagues and accreditation bodies that there is a commitment to quality

Preparation Note

Description

  • This sample is ready to use.
  • Report ′Free Liquid′ in mL of liquid, or as "0", indicating an absence of liquid, or "1", indicating liquid is present.

Storage and Stability

Please refer to sample label for expiration date, specific storage temperature, and specific storage conditions.

Other Notes

Are you new to Supelco® Proficiency Testing? Be sure to register your account details in the PT Portal before placing your order.
This will provide access to data entry, reporting packets and final reports. Registrations will be approved shortly after your order is placed. For further questions, write to us at ptservice@milliporesigma.com.

When placing your order online, please indicate your study preference under the ′your reference′ column. Available studies can be viewed at the bottom of the page.
Click here to learn more about Quick-Turn Studies (on-demand proficiency tests that are offered depending on lot availability). The Quick-Turn results are released within 48-72 hours whereas scheduled study reports are released within 21 days.

Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period concludes.

Proficiency Testing Schedule (2021-2022)


Soils
LPTP21-S2April 7 - May 21, 2021

LPTP22-S2April 6 - May 20, 2022

Legal Information

Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Free liquid .001-100 mL

storage_class_code

13 - Non Combustible Solids

WGK Germany

WGK 3

Flash Point F

Not applicable

Flash Point C

Not applicable

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service