SPE016TIS

Supelco

Dioxins and Furans in Tissue - PT

Proficiency Testing Material

Quality Level

packaging

pkg of 1 × 10 g

matrix

fish (dried)

Featured Industry

Environmental

format

matrix material

General description

Dioxins and Furans in Tissue proficiency testing material is produced following the ISO/IEC 17043:2010, thereby allowing our test components to offer significant performance data to monitor and prove competency to customers, accreditation bodies or both.

Supelco® is a global proficiency testing (PT) provider that manufactures best-in-class environmental and pharmaceutical PT products. We are the world leader in the manufacturing of real-world soil and sediment PT products.

We provide organic PT materials such as BNAs, BTEX/MTBE, diesel, dioxin and furans, gasoline, herbicides, nitrosamines/nitroaromatics, oil and grease, PAHs, PCBs, phenolics, total petroleum hydrocarbons (TPH) and metals in soil, dioxins and furans in tissue, free liquids in paint, PBDE/PCBs in sediment, PCB congeners in fish tissue and toxicity characteristic leaching procedure (TCLP) semi-VOAs.

Type of PT Scheme: Solids/Soil hazardous waste organics

Matrix:
Fish (dried)

Analyte/s: Values of analytes vary from lot to lot.

All information regarding the use of Dioxins and Furans in Tissue Proficiency Testing standard can be found in the reporting packet which will be available to download at PT Portal.

Application

Supelco® Analytical Prep system design offers:

  • reduced solvent usage
  • decrease in sample preparation time (by two days relative to conventional methods)
  • extraction recoveries greater than 85%

Have any questions? Explore our FAQs.
For more information on this product, PT Service Team.

Features and Benefits

We have a vast portfolio of environmental PT schemes with "fit-for-purpose" samples to help you meet the needs of the laboratory.

Our proficiency testing (PT) program is a benchmark to:

  • provide staff with an insight into their laboratory′s performance, to verify the accuracy and reliability of their testing process
  • provide a comparison of performance with that of other laboratories, nationally and internationally
  • test and educate staff, provide a better understanding of the impact of incorrect results and identify areas where there may be problems
  • provide an independent evaluation of laboratory performance
  • demonstrate to clients, colleagues and accreditation bodies that there is a commitment to quality

Preparation Note

Description

  • The PT sample size provided is 10 g of biological fish matrix.
  • Recommended storage condition is 4 ºC.

Sample Preparation

  • For Method 8280A, a 10 g sample size contains sufficient concentrations of analytes such that levels above the method specified quantitative limits will be realized.
  • For Method 8290, a one-gram sample will contain concentrations of analytes such that levels above the method specified quantitative limits will be realized.
  • This holds true though levels of detection will be elevated owing to the reduced sample size used relative to that specified in the method.
  • Report on as received basis.

NOTE: Minimize exposure to the environment, cap when not in use.

Storage and Stability

Please refer to sample label for expiration date, specific storage temperature, and specific storage conditions.

Other Notes

Are you new to Supelco® Proficiency Testing? Be sure to register your account details in the PT Portal before placing your order.
This will provide access to data entry, reporting packets and final reports. Registrations will be approved shortly after your order is placed. For further questions, write to us at ptservice@milliporesigma.com.

When placing your order online, please indicate your study preference under the ′your reference′ column. Available studies can be viewed at the bottom of the page.
Click here to learn more about Quick-Turn Studies (on-demand proficiency tests that are offered depending on lot availability). The Quick-Turn results are released within 48-72 hours whereas scheduled study reports are released within 21 days.

Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period concludes.

Proficiency Testing Schedule (2021-2022)

Soils
LPTP21-S2April 7 - May 21, 2021
LPTP21-S4 October 6 - November 19, 2021

LPTP22-S2April 6 - May 20, 2022
LPTP22-S4 October 5 - November 18, 2022

Legal Information

Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    1,2,3,4,6,7,8-Hpcdd 5-1000 pg/g
    1,2,3,4,6,7,8-Hpcdf 1 5-1000 pg/g
    1,2,3,4,7,8,9-Hpcdf 5-1000 pg/g
    1,2,3,4,7,8-Hxcdd 5-1000 pg/g
    1,2,3,6,7,8-Hxcdd 5-1000 pg/g
    1,2,3,7,8,9-Hxcdd 5-1000 pg/g
    Hxcdd, total 5-5000 pg/g
    1,2,3,4,7,8-Hxcdf 5-1000 pg/g
    1,2,3,6,7,8-Hxcdf 5-1000 pg/g
    1,2,3,7,8,9-Hxcdf 5-1000 pg/g
    2,3,4,6,7,8-Hxcdf 5-1000 pg/g
    1,2,3,4,6,7,8,9-OCDF 5-1000 pg/g
    1,2,3,4,6,7,8,9-OCDD 5-1000 pg/g
    PCDD, total 5-10000 pg/g
    PCDF, total 5-10000 pg/g
    1,2,3,7,8-Pecdd 5-1000 pg/g
    1,2,3,7,8-Pecdf 5-1000 pg/g
    2,3,4,7,8-Pecdf 5-1000 pg/g
    Pecdf, total 5-5000 pg/g
    Pecdd, total 5-5000 pg/g
    TCDD, total 5-5000 pg/g
    2,3,7,8-TCDF 5-1000 pg/g
    TCDF, total 5-5000 pg/g
    2,3,7,8-TCDD 5-1000 pg/g
    Total Heptachlorodibenzo-p-dioxin (Total HPCDD) 5-5000 pg/g
    Total Heptachlorodibenzofuran (Total HPCDF) 5-5000 pg/g
    Total Hexachlorodibenzofuran (Total HxCDF) 5-5000 pg/g
    See All (27)

pictograms

Environment

signalword

Warning

hcodes

pcodes

hazcat

Aquatic Chronic 1

storage_class_code

11 - Combustible Solids

WGK Germany

WGK 3

Flash Point F

Not applicable

Flash Point C

Not applicable

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service