PE1496

Supelco

Cyanide, Total - WS

Proficiency Testing Material

Synonym(s):
WS - Cyanide
NACRES:
NA.24

Quality Level

packaging

ampule of 2 × 2 mL

concentration

in water

Featured Industry

Environmental

format

matrix material

General description

Cyanide, Total - WS proficiency testing material is produced in accordance with ISO/IEC 17043:2010. The PT portfolio allows our test components to offer significant performance data to monitor and prove competency to customers, accreditation bodies or both.

Supelco® is a global proficiency testing (PT) provider that manufactures best-in-class environmental and pharmaceutical PT products. Most water PT samples are available as concentrates or ready-to-use volumes giving the end-user flexibility in selecting samples that will fit their exact needs. Our WS program is designed to meet the quality assurance requirements in compliance with the Safe Drinking Water Act (SDWA), thereby ensuring the analytical competence of your laboratory or drinking water facility.
These samples are typically provided in duplicate for use in case of lab mishap, for QC purposes or to analyze multiple methods.

Type of PT Scheme: Water analysis - Water Supply (WS)

Matrix: Water Supply/Drinking water

Analyte/s: Values of analytes vary lot to lot.

All information regarding the use of Cyanide, Total - WS standard can be found in the reporting packet which will be available to download at PT Portal.

Application

Cyanide, Total - WS proficiency testing material is suitable for testing the cyanide in water supply/drinking water.

Have any questions? Explore our FAQs.
For more information on this product, please contact our PT Service Team.

Features and Benefits

We have a vast portfolio of environmental PT schemes with "fit-for-purpose" samples to help you meet the needs of the laboratory.

Our proficiency testing (PT) program is a benchmark to:

  • provide staff with an insight into their laboratory′s performance, to verify the accuracy and reliability of their testing process
  • provide a comparison of performance with that of other laboratories, nationally and internationally
  • test and educate staff, provide a better understanding of the impact of incorrect results and identify areas where there may be problems
  • provide an independent evaluation of laboratory performance
  • demonstrate to clients, colleagues and accreditation bodies that there is a commitment to quality

Preparation Note

Description

  • This PT sample is as a 2 mL concentrate.
  • The solvent for this sample is 1% NaOH in water.
  • The PT sample has not been preserved.

Sample Preparation


  • Partially fill a 1000 mL volumetric flask with reagent-grade water (near 20 °C) and add 1 mL of 1 N NaOH.
  • Open ampule PE1496-2ML (be careful to avoid cuts on glass) and transfer 1.0 mL of concentrate to the volumetric flask.
  • Fill the volumetric flask to volume with reagent-grade water and mix well.
  • The sample is now ready for immediate analysis.
  • A blank reagent-grade water sample should be analyzed concurrently for background correction. Remember to match the NaOH matrix of the sample.

NOTES: Keep the ampule shielded from ultraviolet radiation. Direct exposure to sunlight or any source of UV radiation may cause a significant change in concentration. The specified addition of 1 mL of 1 N NaOH is only a recommended minimum to prevent the loss of cyanide. If another amount or even another base (such as KOH) is part of the analyst′s normal routine, it is his/her option to change the recommendation.

Storage and Stability

Please refer to sample label for expiration date, specific storage temperature, and specific storage conditions.

Other Notes

Are you new to Supelco® Proficiency Testing? Be sure to register your account details in the PT Portal before placing your order.
This will provide access to data entry, reporting packets and final reports. Registrations will be approved shortly after your order is placed. For further questions, write to us at ptservice@milliporesigma.com.

When placing your order online, please indicate your study preference under the ′your reference′ column. Available studies can be viewed at the bottom of the page.
Click here to learn more about Quick-Turn Studies (on-demand proficiency tests that are offered depending on lot availability). The Quick-Turn results are released within 48-72 hours whereas scheduled study reports are released within 21 days.

Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period is concludes.

Proficiency Testing Schedule (2021-2022)


Drinking Water
WS21-2March 24 - May 7, 2021
WS21-4 September 22 - November 5, 2021

WS22-2March 23 - May 6, 2022
WS22-4 September 21 - November 4, 2022

Legal Information

Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Total cyanide 0.100 - 1.00

storage_class_code

10 - Combustible liquids

WGK Germany

WGK 1

Flash Point F

Not applicable

Flash Point C

Not applicable

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service