Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

792993

Sigma-Aldrich

Specially Denatured Alcohol

190 proof, SDA 3A, contains Methanol

Synonym(s):

Ethanol, SDA

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

CAS Number:
MDL number:
UNSPSC Code:
15101802
PubChem Substance ID:

application(s):
life science and biopharma

form

liquid

contains

Methanol

concentration

190 proof

application(s)

life science and biopharma

Tax and license information

Permit for use required

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

5GA is in metal drum with flex spout.
5GA-PB is in a poly drum with flex spout.
3x5GA is in metal drum with flex spout.
3x5GA-PB is in a poly drum with flex spout.
55GA is in metal drum.
55GA-PB is in poly drum.

Components

95.24% (v/v) Ethanol, 190 Proof
4.76% (v/v) Methanol

Other Notes

Ordering Information:
To purchase Specially Denatured Alcohol, customers must possess an Industrial Alcohol User Permit from the Alcohol and Tobacco Tax and Trade Bureau (TTB).
  • Permit must have customer signature and date of submission to Sigma-Aldrich in addition to the original signature and date.
  • Permit must have word, "copy" handwritten or stamped.
  • Permit number must contain correct state or US designation matching ship-to address.
  • Permit address must match ship-to-address in Sigma-Aldrich system.

TTB Link

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 2

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

60.8 °F - closed cup

Flash Point(C)

16 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Alcohols
Hazardous rank II

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

792993-5X5GA:
792993-5X5GA-PB:
792993-3X5GA-PB:
792993-3X5GA:
792993-55GA:
792993-5GA:
792993-55GA-PB:
792993-5GA-PB:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

U Allenstein et al.
Soft matter, 11(25), 5053-5059 (2015-06-02)
Cells require adhesion to survive, proliferate and migrate, as well as for wound healing and many other functions. The strength of contractile cell forces on an underlying surface is a highly relevant quantity to measure the affinity of cells to
Sonia Soloneski et al.
Toxicology in vitro : an international journal published in association with BIBRA, 29(5), 834-844 (2015-03-31)
In vitro effects of the carbamates pirimicarb and zineb and their formulations Aficida® (50% pirimicarb) and Azzurro® (70% zineb), respectively, were evaluated in Chinese hamster ovary (CHO-K1) cells. Whereas the cytokinesis-blocked micronucleus cytome assay was employed to test for genotoxicity
Nicholas J Niemuth et al.
Chemosphere, 135, 38-45 (2015-04-22)
The occurrence of intersex fish, where male reproductive tissues show evidence of feminization, have been found in freshwater systems around the world, indicating the potential for significant endocrine disruption across species in the ecosystem. Estrogens from birth control medications in
Danielle Hagstrom et al.
Toxicological sciences : an official journal of the Society of Toxicology, 147(1), 270-285 (2015-06-28)
Traditional toxicology testing has relied on low-throughput, expensive mammalian studies; however, timely testing of the large number of environmental toxicants requires new in vitro and in vivo platforms for inexpensive medium- to high-throughput screening. Herein, we describe the suitability of the asexual

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service