Merck
All Photos(1)

32274

Supelco

4-(3,6-Dimethyl-3-heptyl)phenol-3,5-d2 solution

100 μg/mL in acetone, analytical standard

Synonym(s):
3,6,3-Nonylphenol-d2, 363-NP-D2, 4-(1-Ethyl-1,4-dimethylpentyl)phenol-3,5-d2
Empirical Formula (Hill Notation):
C15D2H22O
CAS Number:
Molecular Weight:
222.36
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetone

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

single component solution

mass shift

M+2

storage temp.

−20°C

SMILES string

[2H]c1cc(O)cc([2H])c1C(C)(CC)CCC(C)C

InChI

1S/C15H24O/c1-5-15(4,11-10-12(2)3)13-6-8-14(16)9-7-13/h6-9,12,16H,5,10-11H2,1-4H3/i6D,7D

InChI key

BGLUSMSXKMBLFS-QFIQSOQBSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.0 °C - closed cup

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service