Merck
All Photos(1)

15-0020

Sigma-Aldrich

Imidazole

SAJ special grade, ≥99.0%

Synonym(s):
1,3-Diaza-2,4-cyclopentadiene, Glyoxaline
Empirical Formula (Hill Notation):
C3H4N2
CAS Number:
Molecular Weight:
68.08
Beilstein:
103853
EC Number:
MDL number:
PubChem Substance ID:

grade

SAJ special grade

vapor pressure

<1 mmHg ( 20 °C)

assay

≥99.0%

form

crystalline

availability

available only in Japan

pH

9-11 (23 °C, 100 g/L)

pKa (25 °C)

6.95

bp

256 °C (lit.)

mp

88-91 °C (lit.)

SMILES string

c1c[nH]cn1

InChI

1S/C3H4N2/c1-2-5-3-4-1/h1-3H,(H,4,5)

InChI key

RAXXELZNTBOGNW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Excellent for buffers in the range of pH 6.2-7.8

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 1B - Skin Corr. 1C

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

293.0 °F - closed cup

Flash Point(C)

145 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service