Merck
All Photos(4)

ZRDSVP3WW

Milli-Q

RiOs-DI® Water Purification System

built-in bactericidal UV lamp, A basic solution for Type 2 pure water production with integrated 6 L storage tank

Synonym(s):
RiOs-DI 3 UV Water Purification System

For more information on this product, including pricing & availability, please contact our Technical Service Team

AC/DC input

100 - 230 V AC, 50 - 60 Hz (per your country requirements)

packaging

pack of 1 unit

parameter

17.3 kg operating weight (38.0 lb)
3 L/hr flow rate
40 L/day max. usage (water)

resistivity

>10 MΩ-cm, at 25°C

H × W × D

49.7 cm (19.6 in.) × 29 cm (11.4 in.) × 33.9 cm (13.3 in.)

tank volume

6 L , Integrated storage tank

input

feed water nature potable tap water

output

product water quality: type 2 water (> 5 MΩ·cm)

Looking for similar products? Visit Product Comparison Guide

General description

With a combination of reverse osmosis (RO) technology and deionization (DI) resins, the compact, user-friendly RiOs-DI® 3 UV Water Purification Systems produce up to 15 L per day of high-quality Type 2 pure water—easily and economically.
The combined power of RO and DI resins provides pure water with high resistivity (> 10 MΩ·cm) and low TOC (< 30 ppb) whose quality surpasses that of service DI or distilled water. RiOs-DI® systems let you avoid the many drawbacks of these other purification methods: high lab space demands to store resin bottles or accommodate distillation equipment; heavy maintenance; lack of water quality monitoring; and in the case of distillation, high energy and water use, as well as the need for strong chemicals for cleaning.

Application

RiOs-DI® systems are a source of pure (type 2) water for general laboratory tasks, preparation of buffers, microbiological culture media and reagent solutions, rinsing of laboratory glassware.

The RiOs-DI® 3 UV Water Purification System has a built-in 254 nm UV lamp to eliminate bacteria for critical applications.

Features and Benefits

  • Superior quality pure water: Easy-to-use RiOs-DI® systems produce Type 2 pure water with quality superior to that of service DI or stills. With their combination of reverse osmosis (RO) technology and deionization resins, RiOs-DI® systems let you avoid the issues associated with service DI or distillation (maintenance, storage, cleaning, etc.).
  • Pure water for general laboratory use: Water from RiOs-DI® systems is suitable for a variety of uses, including buffer and reagent preparation; microbiological culture media preparation; and glassware rinsing.
  • Compact design for the most efficient use of your lab space: A small footprint makes it easy to install the RiOs-DI® system wherever you want to in your lab.
  • Flow rate and storage volume to meet your low-volume pure water needs: Systems have a pure water flow rate of >2.4 L per hour and an integrated 6 L reservoir for pure water storage.
  • High-quality water for applications requiring low bacterial levels: A built-in bactericidal UV lamp helps ensure optimal water quality for bacteria-sensitive applications.
  • Easily accessible information on system operation: The user-friendly display provides system status at a glance; the concise Quick Reference Guide is a handy guide for daily operation.
  • Easy installation: The system is designed for easy installation. Just connect the system to a tap water supply, plug it in, and insert the SmartPak® RODI cartridge -your system is ready to use!
  • Simple self-maintenance: The single SmartPak® RODI cartridge enables easy and rapid replacement.

Components

  • This is a web reference. The end letters "WW" will be replaced by the country letters on your invoice to have the right shipping kit for the country. This ensures you receive the user manual in local language and a power cord adapted to the local electrical network.
  • The indicated price currently corresponds to the above catalogue number only; it is subject to change at any time without notice.
  • This catalogue number does not include consumables, accessories and other products and/or services that may be required for the installation and the correct operation of the water purification unit over time.
  • Because our customers have specific purified water needs, we recommend that you have your purchase validated by one of our specialists. We remain at your disposal to advise you; do not hesitate to contact us!

Other Notes

For more information on this product please contact our Technical Service Team

Legal Information

RIOS-DI is a registered trademark of Merck KGaA, Darmstadt, Germany
SMARTPAK is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Quality

Enter Lot Number to search for Certificate of Quality (COQ).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service