Merck
All Photos(1)

XX1504732

Millipore

Glass Base & Cap with stainless steel screen, 47 mm

Accessories for filter holders for sample preparation.

eCl@ss:
32031613

material

borosilicate glass base
stainless steel screen
tubulated borosilicate glass cap

diam.

47 mm

filtration area

9.6 cm2

fitting

6 mm (1/4 in.) O.D. outlet tubulated cap sidearm for vacuum

application(s)

filtration

shipped in

ambient

General description

REMOVE FROM SCOPE

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Quality

Enter Lot Number to search for Certificate of Quality (COQ).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service