Merck
All Photos(1)

MXHVWP124

Millipore

Milliflex® funnel

100mL, 0.45μm white plain Durapore® membrane for bioburden and water analysis with the Milliflex® system

NACRES:
NB.24

material

PVDF (hydrophilic)
plain filter
polypropylene
styrene-acrylonitrile (SAN)
white filter

Quality Level

sterility

sterile; ethylene oxide treated

manufacturer/tradename

Milliflex®

parameter

100 mL sample volume

filter diam.

51 mm

matrix

Durapore®

pore size

0.45 μm pore size

input

sample type bottled water
sample type cosmetics
sample type pharmaceutical(s)
sample type beer

application(s)

bioburden testing
cosmetics
pharmaceutical
water monitoring

shipped in

ambient

Related Categories

General description

Milliflex® funnel is a ready-to-use, sterilized filter unit that combines a 100mL funnel and a membrane filter in a single device offering a complete solution for bioburden testing. The funnel unit is made of polypropylene, SAN and the membrane is made of white plain, 51mm diameter polyvinylidene difluoride (Durapore®) with 0.45μm pore size. The sealed integral membrane filter eliminates the need for membrane manipulation and handling, thus minimizing a major source of contamination and thereby improves bioburden testing reliability. The sealed membrane prevents the bypass phenomenon often found with other filtration systems. The Milliflex® filtration possesses almost twice the surface area of standard membranes enabling faster filtration and the funnel can be easily detached from the membrane without the use of any tools. The convex shape of the membrane after filtration demonstrates the integrity of the membrane and prevents air bubbles from being trapped when placed on culture media.

Application

Milliflex® 100mL funnel with Durapore® membrane is used in Bioburden testing, water monitoring and general microbiological analysis of bottled water and beer. It is widely used in pharmaceutical, cosmetic and brewing industry

Legal Information

Durapore is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Quality

Enter Lot Number to search for Certificate of Quality (COQ).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service