T-163

Supelco

2-Thiofuranyl fentanyl hydrochloride solution

new

100 μg/mL in methanol ((as free base)), certified reference material, ampule of 0.5 mL, Cerilliant®

Empirical Formula (Hill Notation):
C24H26N2OS · HCl
CAS Number:
Molecular Weight:
427.00

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 0.5 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol ((as free base))

format

single component solution

storage temp.

−20°C

SMILES string

O=C(C1=CC=CS1)N(C2CCN(CCC3=CC=CC=C3)CC2)C4=CC=CC=C4.Cl

General description

This Certified Spiking Solution® is suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis, clinical toxicology to urine drug testing. 2-Thiofuranyl fentanyl is an opioid analgesic that is an analog of fentanyl and has been sold as a designer drug.

Legal Information

CERILLIANT is a registered trademark of Cerilliant Corporation
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Cerilliant Corporation

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service