HomeWebinarsAddressing Immunoglobulin (Ig) Purification Challenges with Chromatographic Technologies
Addressing Immunoglobulin (Ig) Purification Challenges with Chromatographic Technologies
Webinar
Abstract - A webinar to discuss strategies to address Immunoglobulin (Ig) purification challenges by using Ion Exchange and Affinity chromatography media.The plasma industry is facing pressure to provide a safe, efficacious and secure supply of plasma products around the globe. There is a particular focus on the top leading product of this industry, Immunoglobulin. From a manufacturing perspective, many efforts have been set on increasing yield and safety using chromatography steps.
In this webinar you will learn about how our Fractogel® EMD Ion-Exchange and Eshmuno® P anti-A/B affinity chromatography resins can support your purification goals by
  • Enhancing yield and purity
  • Providing long and robust lifecycles Fractogel® EMD TMEA is an anion exchange chromatography resin, using our proprietary tentacle technology to effectively increase target protein binding capacity. Eshmuno® P anti-A and Eshmuno® P anti-B are two distinct affinity-based chromatography resins specifically designed to effectively remove anti-A and anti-B isoagglutinins, respectively
Speakers
Eric Youssef
Merck
Associate Director Plasma Initiative
Webinar Information
Pharmaceutical and biopharmaceutical manufacturing
  • Plasma manufacturing
  • Duration:53 min
  • Language:English
  • Session 1:Wed, August 18, 2021
    09:38 AM CST
  • Session 2:Mon, September 13, 2021
    05:38 PM CST