Merck
HomeWebinarsNGSO (Next-Gen Sequencing Oligos) for Consistent and Proper Sequence Assembly
NGSO (Next-Gen Sequencing Oligos) for Consistent and Proper Sequence Assembly
Webinar
Optimized NGS performance relies on oligo adapter quality. Adapters – especially index adapters since they contain the multiplex identifier or barcode – containing too high a proportion of truncated sequences (unacceptably low purity) or too high a proportion of other adapter sequences (excessive cross contamination) can lead to compromised sequence read integrity and improper sequence assembly, respectively, during multiplex NGS experiments. These types of problems are undetectable until the data analysis stage, which make them costly in terms of time and money. Therefore, the production process used for the adapter sequences is critical for a successful sequencing run. NGSO are manufactured under rigorous conditions to ensure suitable purity and low cross contamination to meet research, commercial, and molecular diagnostic needs.
Speaker
Tom Russell, Ph.D
Merck
Product Manager, Custom Products
Webinar Information
Clinical testing and diagnostics manufacturing
Microbiological testing
  • Duration:1 hour
  • Language:English
  • Presented:June 16, 2021