Merck
PTFE Packed GC Column (General Configuration) phase none, matrix Chromosil 310 support, L × O.D. × I.D. 8.0 ft (2.4 m) × 1/8 in. × 2.1 mm, Ends empty; middle 6 ft packed
PTFE Packed GC Column (General Configuration)
PTFE Packed GC Column (General Configuration) phase none, matrix Chromosil 310 support, L × O.D. × I.D. 8.0 ft (2.4 m) × 1/8 in. × 2.1 mm, Ends empty; middle 6 ft packed
Products
Categoria del prodotto
Marchio
Tipo di colonna
ID (mm)
Lunghezza (m)