Documenti fondamentali
Z707643
TPP® cell culture spatula
Sinonimo/i:
cell scrapers, cell spatula
About This Item
Codice UNSPSC:
41122103
Prezzi e disponibilità al momento non sono disponibili
Materiali
polypropylene
Sterilità
sterile
Confezionamento
pack of 1
case of 150
Produttore/marchio commerciale
TPP 99010
Pala lung × larg
195 mm × 14 mm , fixed blade
Misura
Compatibilità
suitable for (tissue culture dish 150, various other cell culture applications)
Confronta articoli simili
Visualizza il confronto per intero
Mostra le differenze
1 of 4
Questo articolo
HPA010698HPA018674HPA012945
antibody form
affinity isolated antibody
antibody form
affinity isolated antibody
antibody form
affinity isolated antibody
antibody form
affinity isolated antibody
conjugate
unconjugated
conjugate
unconjugated
conjugate
unconjugated
conjugate
unconjugated
biological source
rabbit
biological source
rabbit
biological source
rabbit
biological source
rabbit
product line
Prestige Antibodies® Powered by Atlas Antibodies
product line
Prestige Antibodies® Powered by Atlas Antibodies
product line
Prestige Antibodies® Powered by Atlas Antibodies
product line
Prestige Antibodies® Powered by Atlas Antibodies
form
buffered aqueous glycerol solution
form
buffered aqueous glycerol solution
form
buffered aqueous glycerol solution
form
buffered aqueous glycerol solution
Descrizione generale
The TPP cell spatula is mainly used with TPP tissue culture flasks with re-closable lid or peel off foil and with the large TPP tissue culture dish.
The advantages are:
  • Special formed blade with sharp edges
  • Small raised knobs on the handle
  • Lint-free blister packaging
  • User friendly TPP dispenser case
Note legali
TPP is a registered trademark of TPP Techno Plastic Products AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)