Documenti fondamentali
DWK209043
Wheaton® Leak Resistant Bottle
capacity 8 mL, high-density polyethylene bottle, natural bottle, narrow-mouth bottle, bottle diam. × H 25 mm × 43 mm, 20-410
Sinonimo/i:
209043
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
Materiali
high-density polyethylene bottle
narrow-mouth bottle
natural bottle
Confezionamento
pkg of 72 ea
Produttore/marchio commerciale
WHEATON® 209043
Diametro bottiglia × alt.
25 mm × 43 mm
Capacità
8 mL
Caratteristiche e vantaggi
Superior double seal closure for leak resistance
Note legali
WHEATON is a registered trademark of Wheaton Industries, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)