Merck
  • Single cell analysis of phosphoinositide 3-kinase/Akt and ERK activation in acute myeloid leukemia by flow cytometry.
Single cell analysis of phosphoinositide 3-kinase/Akt and ERK activation in acute myeloid leukemia by flow cytometry.
Haematologica (2006-06-14)
Valérie Bardet, Jerome Tamburini, Norbert Ifrah, François Dreyfus, Patrick Mayeux, Didier Bouscary, Catherine Lacombe
ABSTRACT
Abnormal activation of several signal transduction pathways such as phosphoinositide 3-kinase (PI3K) and MAP kinases has been reported in acute myeloid leukemia (AML). To test new targeted therapeutics, it is critical to develop sensitive analytical tools to detect abnormal activation of these pathways and to monitor their inhibition in response to treatment. We analyzed Akt and ERK phosphorylation in 32 samples from patients using western blot and a two-color flow cytometry protocol using CD34. To circumvent the CD34 negative AML found in our series, we developed a two-color protocol using CD45 to isolate the blast cell population. Finally, a four-color protocol was used to detect phosphorylation in an enriched population of AML stem cells. We compared western blot analysis and flow cytometry for the detection of PI3K/Akt and ERK activation and found a 100% correlation between the two techniques in a series of 32 AML samples. Using a flow cytometry protocol, we were able to analyze all the patients' samples, even those with low blast infiltration or CD34 negative blast cells. We were also able to detect the phosphorylated proteins in the most immature blast cell population with the CD34+ CD38-/low CD123+ phenotype. Interpretations and Our study shows that flow cytometry is a reliable method for detecting Akt and ERK phosphorylation in all patients' samples. Activation can also be detected in the most immature blast cells, which represent exquisite target cells for new therapeutics.
Sigma-Aldrich
IgG from rabbit serum, technical grade, ≥80% (SDS-PAGE), buffered aqueous solution