Merck
  • Alzheimer amyloid beta inhibition of Eg5/kinesin 5 reduces neurotrophin and/or transmitter receptor function.
Alzheimer amyloid beta inhibition of Eg5/kinesin 5 reduces neurotrophin and/or transmitter receptor function.
Neurobiology of aging (2014-03-19)
Csilla Ari, Sergiy I Borysov, Jiashin Wu, Jaya Padmanabhan, Huntington Potter
ABSTRACT
The mechanism by which amyloid beta (Aβ) causes neuronal dysfunction and/or death in Alzheimer's disease (AD) is unclear. Previously, we showed that Aβ inhibits several microtubule-dependent kinesin motors essential for mitosis and also present in mature neurons. Here, we show that inhibition of kinesin 5 (Eg5) by Aβ blocks neuronal function by reducing transport of neurotrophin and neurotransmitter receptors to the cell surface. Specifically, cell-surface NGF/NTR(p75) and NMDA receptors decline in cells treated with Aβ or the kinesin 5 inhibitor monastrol, or expressing APP. Aβ and monastrol also inhibit NGF-dependent neurite outgrowth from PC12 cells and glutamate-dependent Ca++ entry into primary neurons. Like Aβ, monastrol inhibits long-term potentiation, a cellular model of NMDA-dependent learning and memory, and kinesin 5 activity is absent from APP/PS transgenic mice brain or neurons treated with Aβ. These data imply that cognitive deficits in AD may derive in part from inhibition of neuronal Eg5 by Aβ, resulting in impaired neuronal function and/or survival through receptor mislocalization. Preventing inhibition of Eg5 or other motors by Aβ may represent a novel approach to AD therapy.
Sigma-Aldrich
N-Methyl-D-aspartic acid, ≥98% (TLC), solid