- Mid-IR supercontinuum generation in a ZBLAN fiber pumped by a gain-switched mode-locked Tm-doped fiber laser and amplifier system.
Mid-IR supercontinuum generation in a ZBLAN fiber pumped by a gain-switched mode-locked Tm-doped fiber laser and amplifier system.
Optics express (2013-04-11)
Jacek Swiderski, Maria Michalska, Gwenael Maze
PMID23571875
ABSTRACT
We demonstrate a novel method of mid-infrared (mid-IR) supercontinuum (SC) generation with the use of a 2 µm gain-switched self-mode-locked thulium-doped fiber laser. SC radiation ranging from ~1.9 to 3.8 µm wavelength, generated in a single-mode ZBLAN fiber with a zero-dispersion wavelength (ZDW) shifted to ~1.9 µm, is reported. An average output power of 0.74 W with 0.27 W at wavelengths longer than 2.4 µm was measured. It is, to the best of our knowledge, the first report on such an approach to generate a mid-IR SC in optical fibers.