Merck
  • Neuropharmacological screening of two 1,5-benzodiazepine compounds in mice.
Neuropharmacological screening of two 1,5-benzodiazepine compounds in mice.
Comptes rendus biologies (2010-03-27)
Wafa Ben-Cherif, Rafik Gharbi, Hichem Sebai, Dorra Dridi, Naceur A Boughattas, Mossadok Ben-Attia
ABSTRACT
This work investigates whether the two 1,5-benzodiazepine compounds: 4-(2-hydroxyphenyl)-1,5-benzodiazepin-2-one (RG0501) and Benzopyrano [4,3-c] 1,5-benzodiazepine (RG0502) have any neuropharmacological activities. Diazepam and Flunitrazepam were used as drug references. The investigational 1,5-BDZ were tested in vivo for potentiating hexobarbital-induced sleep and pentylenetetrazole (PTZ)-induced seizures. Our study demonstrated that the increase of sleep duration was significantly higher with RG0501 as compared to RG0502. However, RG0502 anticonvulsant effect was more pronounced than that of RG0501 in the range dose of 6.25-37.5 mg.kg(-1). From the 50 mg.kg(-1) dose, RG0502 offered a protection against clonic-tonic seizures as well as lethality (p< or =0.05). The results showed that the required doses to obtain a pharmacological activity were more than those of the references. This difference could be related to the lack of specific substituants responsible for the pharmacological activity in the tested compounds.
Product Description
Supelco
Hexobarbital solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®