Merck
  • Inactivation of prolyl hydroxylase domain (PHD) protein by epigallocatechin (EGCG) stabilizes hypoxia-inducible factor (HIF-1α) and induces hepcidin (Hamp) in rat kidney.
Inactivation of prolyl hydroxylase domain (PHD) protein by epigallocatechin (EGCG) stabilizes hypoxia-inducible factor (HIF-1α) and induces hepcidin (Hamp) in rat kidney.
Biochemical and biophysical research communications (2011-12-06)
Dominador J Manalo, Jin Hyen Baek, Paul W Buehler, Evi Struble, Bindu Abraham, Abdu I Alayash
ABSTRACT
HIF-1α plays a key role in iron uptake and transport in the liver, whose activity is tightly linked to the repression of hepcidin (Hamp). Hamp prevents intestinal iron uptake and cellular efflux by negatively modulating ferroportin. Hamp is also expressed in the kidneys, where transcriptional control by HIF-1α remains poorly understood. We show that the administration of epigallocatechin gallate (EGCG) results in a considerable Hamp expression in rat kidneys. We also provide evidence to show that EGCG inhibited prolyl hydroxylase (PHD) activity, essential for HIF-1α degradation in vivo and in vitro. Rats that were dosed with EGCG (60 mg/kg, intraperitoneal) over a 7 day time course stabilized HIF-1α protein in kidney tissues. Interestingly, Hamp gene expression was induced, even after subjecting rats to a 4h hypoxia treatment (8% oxygen). Using Hep3B cells, we determined that EGCG conferred its inhibitory action by complexing with PHD, altering its catalytic iron center and thus preventing HIF-1α hydroxylation. These data demonstrate EGCG's therapeutic potential in modulating hepcidin expression in diseases associated with altered iron metabolism.
Sigma-Aldrich
(−)-Gallocatechin, ≥98% (HPLC)
Supelco
(−)-Gallocatechin, analytical standard