- Vascular protective role of vitexicarpin isolated from Vitex rotundifolia in human umbilical vein endothelial cells.
Vascular protective role of vitexicarpin isolated from Vitex rotundifolia in human umbilical vein endothelial cells.
Pro-inflammatory cytokines induce injury of endothelial cells caused by increases of adhesion molecules, leading to vascular inflammation and the development of atherosclerosis. Recent pharmacological studies have demonstrated that vitexicarpin, a flavonoid isolated from Vitex rotundifolia, has anti-inflammatory, antitumor, and analgesic properties. In this study, we investigated whether vitexicarpin (5-100 nM) prevented the TNF-α-induced vascular inflammation process in human umbilical vein endothelial cells (HUVEC). We found that pretreatment with vitexicarpin decreased TNF-α (10 ng/ml)-induced expression of cell adhesion molecules such as vascular cell adhesion molecule-1, intracellular adhesion molecule-1, and E-selectin as well as matrix metalloproteinase-2 and -9 expression. Preincubation with vitexicarpin also dose-dependently inhibited TNF-α-induced adhesion of HL-60 monocytic cells. Vitexicarpin significantly decreased TNF-α-induced intracellular reactive oxygen species (ROS) production. Furthermore, vitexicarpin suppressed NF-κB nuclear translocation and transcriptional activity in TNF-α-treated HUVEC. In conclusion, vitexicarpin significantly reduced vascular inflammation, through inhibition of ROS-NF-κB pathway in vascular endothelial cells.